Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 7 | 8334 | ||
NM_018299.4 | NP_060769.4 | 6 | 8301 | |||
NR_073119.1 | 6 | 8322 | ||||
NR_073120.1 | 6 | 8282 | ||||
NR_073121.1 | 5 | 8198 | ||||
GenBank mRNA | AK123531.1 | - | 1 | 2684 |