Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308213.1 | NP_001295142.1 | 9 | 1262 | ||
NM_001308219.1 | NP_001295148.1 | 5 | 762 | |||
NM_001308223.1 | NP_001295152.1 | 4 | 659 | |||
XM_005265860.2 | XP_005265917.1 | 9 | 1255 | |||
XM_011534494.1 | XP_011532796.1 | 8 | 1161 | |||
XM_017009294.1 | XP_016864783.1 | 8 | 1235 | |||
XM_017009295.1 | XP_016864784.1 | 8 | 1187 | |||
GenBank mRNA | AK022292.1 | - | 1 | 945 | ||
AK131469.1 | - | 10 | 1207 | |||
AK297584.1 | - | 4 | 643 | |||
BX331331.2 | - | 5 | 717 | |||
CR627467.1 | - | 1 | 112 |