Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005247121.4 | XP_005247178.1 | 38 | 9710 | ||
XM_011512423.2 | XP_011510725.1 | 37 | 9775 | |||
XM_011512424.2 | XP_011510726.1 | 37 | 9775 | |||
XM_011512425.2 | XP_011510727.1 | 39 | 9890 | |||
XM_011512426.2 | XP_011510728.1 | 39 | 9784 | |||
XM_011512428.2 | XP_011510730.1 | 38 | 9811 | |||
XM_017005711.1 | XP_016861200.1 | 40 | 9995 | |||
XM_017005712.1 | XP_016861201.1 | 39 | 10099 | |||
XM_017005713.1 | XP_016861202.1 | 40 | 10046 | |||
XM_017005714.1 | XP_016861203.1 | 38 | 16151 | |||
XM_017005715.1 | XP_016861204.1 | 39 | 9916 | |||
GenBank mRNA | AF086237.1 | - | 1 | 457 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM725130.1 | - | 1 | 733 | Chr. 3 - 130677393 - 130678155 [+] on Build GRCh38 |