Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC283435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038735.1 | - | 3, 4 | 337 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI460416.1 | - | 3, 4 | 342 | Chr. 12 - 126401249 - 126449328 [-] on Build GRCh38 |