Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007202.3 | NP_009133.2 | 15 | 3568 | ||
XM_006721432.3 | XP_006721495.1 | 14 | 3389 |