Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110108.1 | 3 | 329 | |||
NR_110109.1 | 5 | 446 | ||||
NR_110110.1 | 3 | 358 | ||||
NR_110111.1 | 2 | 216 | ||||
GenBank mRNA | AB849958.1 | - | 3 | 329 | ||
AB849959.1 | - | 3 | 329 | |||
AB849960.1 | - | 5 | 446 | |||
AB849961.1 | - | 5 | 446 | |||
AB849962.1 | - | 2 | 216 | |||
AB849963.1 | - | 3 | 358 | |||
AB849964.1 | - | 3 | 358 | |||
AK001551.1 | - | 3 | 329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU130161.1 | - | 3 | 329 | Chr. 12 - 105704203 - 105742411 [+] on Build GRCh38 |