Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110108.1 | 3 | 839 | |||
NR_110109.1 | 5 | 956 | ||||
NR_110110.1 | 3 | 868 | ||||
NR_110111.1 | 2 | 726 | ||||
GenBank mRNA | AB849958.1 | - | 3 | 839 | ||
AB849959.1 | - | 3 | 839 | |||
AB849960.1 | - | 5 | 956 | |||
AB849961.1 | - | 5 | 956 | |||
AB849962.1 | - | 2 | 726 | |||
AB849963.1 | - | 3 | 868 | |||
AB849964.1 | - | 3 | 868 | |||
AK001551.1 | - | 3 | 839 |