Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003818.3 | NP_003809.1 | 13 | 8564 | ||
XM_006723660.1 | XP_006723723.1 | 13 | 8575 | |||
XM_011529390.1 | XP_011527692.1 | 14 | 8887 | |||
GenBank mRNA | AK022998.1 | - | 1 | 1908 | ||
AK123831.1 | - | 1 | 2860 | |||
BC015432.1 | - | 1 | 218 | |||
BC025346.1 | - | 1 | 497 | |||
BC044234.1 | - | 1 | 415 |