Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TFDP2 SIRNA Assays ›
Gene Symbol:
TFDP2
Gene Name:
transcription factor Dp-2
Gene Aliases:
DP2
Chromosome Location:
Chr.3: 141944428 - 142149544 on Build GRCh38
UniGene ID:
Hs.379018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n364921
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TFDP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178138.1
NP_001171609.1
14
7669
NM_001178139.1
NP_001171610.1
13
7631
NM_001178140.1
NP_001171611.1
10
7127
NM_001178141.1
NP_001171612.1
10
7060
NM_001178142.1
NP_001171613.1
8
6908
NM_006286.4
NP_006277.1
11
7185
XM_005247731.2
XP_005247788.1
13
7634
XM_011513101.2
XP_011511403.1
14
7730
XM_011513102.2
XP_011511404.1
12
7303
XM_011513106.2
XP_011511408.1
14
7727
XM_011513108.2
XP_011511410.1
11
7457
XM_011513109.2
XP_011511411.1
9
7029
XM_011513111.2
XP_011511413.1
9
7026
XM_017007091.1
XP_016862580.1
13
7642
XM_017007092.1
XP_016862581.1
16
7888
XM_017007093.1
XP_016862582.1
16
8077
XM_017007094.1
XP_016862583.1
13
7663
XM_017007095.1
XP_016862584.1
15
7853
XM_017007096.1
XP_016862585.1
12
7560
XM_017007097.1
XP_016862586.1
14
7659
XM_017007098.1
XP_016862587.1
14
7807
XM_017007099.1
XP_016862588.1
15
7782
XM_017007101.1
XP_016862590.1
13
7615
XM_017007102.1
XP_016862591.1
11
7410
GenBank mRNA
AK096450.1
-
1
715
BC040991.2
-
1
42
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription cofactor activity
protein binding
transcription factor binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline