Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125887.1 | - | 1 | 167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB068097.1 | - | 1 | 168 | Chr. 9 - 104086497 - 104092474 [-] on Build GRCh38 |