Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712677.3 | XP_006712740.1 | 7 | 5408 | ||
GenBank mRNA | AK055724.1 | - | 1 | 2062 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU191922.1 | - | 1 | 416 | Chr. 2 - 210024057 - 210024609 [+] on Build GRCh38 |