Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033805.1 | 3 | 1112 | |||
GenBank mRNA | AK001143.1 | - | 1 | 323 | ||
BC045818.1 | - | 3 | 1092 |