Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002185.3 | NP_002176.2 | 8 | 4481 | ||
NR_120485.1 | 6 | 4218 | ||||
XM_005248299.3 | XP_005248356.1 | 7 | 4372 | |||
GenBank mRNA | AF373204.1 | - | 1 | 562 | ||
AL713738.1 | - | 1 | 1169 |