Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003807.4 | NP_003798.2 | 5 | 4137 | ||
NM_172014.3 | NP_742011.2 | 4 | 3736 | |||
XM_017027417.1 | XP_016882906.1 | 4 | 4281 | |||
GenBank mRNA | AK026704.1 | - | 1 | 1562 |