Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055490.1 | - | 1 | 219 |
Gene Symbol: LOC100130856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095727.1 | - | 1 | 1951 |