Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524342.2 | XP_011522644.1 | 5 | 6246 | ||
XM_017024192.1 | XP_016879681.1 | 5 | 6250 | |||
XM_017024194.1 | XP_016879683.1 | 5 | 2399 | |||
XM_017024195.1 | XP_016879684.1 | 5 | 2403 | |||
GenBank mRNA | AK054819.1 | - | 1 | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM787371.1 | - | 1 | 23 | Chr. 17 - 54957386 - 54958066 [-] on Build GRCh38 |