Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524342.2 | XP_011522644.1 | 5 | 6001 | ||
XM_017024192.1 | XP_016879681.1 | 5 | 6005 | |||
XM_017024194.1 | XP_016879683.1 | 5 | 2154 | |||
XM_017024195.1 | XP_016879684.1 | 5 | 2158 | |||
GenBank mRNA | AK054819.1 | - | 1 | 138 |