Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317110.1 | NP_001304039.1 | 6 | 3694 | ||
NM_001317111.1 | NP_001304040.1 | 6 | 3590 | |||
NM_001317112.1 | NP_001304041.1 | 5 | 3533 | |||
NM_018221.4 | NP_060691.2 | 6 | 3697 | |||
XM_011532948.2 | XP_011531250.1 | 5 | 3393 | |||
GenBank mRNA | AL122062.1 | - | 1 | 1412 |