Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011519345.2 | XP_011517647.1 | 1 | 549 | ||
GenBank mRNA | AK123512.1 | - | 1 | 501 |