Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029783.1 | - | 1 | 968 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG617189.1 | - | 1 | 170 | Chr. 17 - 42386834 - 42387623 [-] on Build GRCh38 |