Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011532924.2 | XP_011531226.1 | 24 | 5778 | ||
GenBank mRNA | AK027272.2 | - | 1 | 1873 |