Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324242.1 | NP_001311171.1 | 8 | 2834 | ||
NM_001324243.1 | NP_001311172.1 | 8 | 2927 | |||
NM_018301.4 | NP_060771.3 | 7 | 2762 | |||
NR_136728.1 | 7 | 2430 | ||||
NR_136729.1 | 8 | 2524 | ||||
NR_136730.1 | 8 | 3558 | ||||
NR_136732.1 | 7 | 2523 | ||||
XM_011530981.2 | XP_011529283.1 | 7 | 2854 | |||
GenBank mRNA | AL832966.1 | - | 1 | 867 |