Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U61096.1 | - | 1 | 210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG187120.1 | - | 1 | 74 | Chr. 3 - 178525638 - 178526378 [+] on Build GRCh38 |