Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509649.2 | XP_011507951.1 | 1 | 17 | ||
XM_011509650.2 | XP_011507952.1 | 1 | 17 |
Gene Symbol: LOC107983947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 851 |