Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011509644.2 | XP_011507946.1 | 1 | 138 | ||
XM_011509645.2 | XP_011507947.1 | 1 | 135 | |||
XM_011509649.2 | XP_011507951.1 | 1 | 146 | |||
XM_011509650.2 | XP_011507952.1 | 1 | 146 |
Gene Symbol: LOC107983947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510285.1 | XP_011508587.1 | 1 | 980 |