Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510158.2 | XP_011508460.1 | 2 | 5917 | ||
GenBank mRNA | AL050068.1 | - | 1 | 814 |
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW090198.1 | - | 1 | 65 | ||
BM999456.1 | - | 1 | 29 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY245074.1 | - | 1 | 43 | Chr. 1 - 202133408 - 202133799 [+] on Build GRCh38 |