Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR37L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510158.2 | XP_011508460.1 | 2 | 5398 | ||
GenBank mRNA | AL050068.1 | - | 1 | 295 |
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM688638.1 | - | 1 | 88 |