Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110832.1 | 1 | 231 | |||
GenBank mRNA | BC033981.1 | - | 1 | 237 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI559473.1 | - | 236 | Chr. - - [-] on Build |