Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004580.4 | NP_004571.2 | 6 | 3234 | ||
NM_183234.2 | NP_899057.1 | 7 | 3215 | |||
NM_183235.2 | NP_899058.1 | 7 | 3224 | |||
NM_183236.2 | NP_899059.1 | 7 | 3175 | |||
XM_005254576.4 | XP_005254633.1 | 6 | 4477 | |||
XM_011521852.1 | XP_011520154.1 | 9 | 3438 | |||
XM_011521854.1 | XP_011520156.1 | 8 | 3311 | |||
XM_011521855.2 | XP_011520157.1 | 8 | 4700 | |||
XM_011521856.2 | XP_011520158.1 | 6 | 3107 | |||
GenBank mRNA | BC078139.1 | - | 2057 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BP423299.1 | - | 1 | 215 | Chr. 15 - 55202966 - 55203405 [-] on Build GRCh38 |