Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317911.1 | NP_001304840.1 | 4 | 5069 | ||
NM_001317912.1 | NP_001304841.1 | 5 | 5169 | |||
NM_139245.3 | NP_640338.2 | 4 | 5493 | |||
NR_134243.1 | 6 | 5408 | ||||
GenBank mRNA | AK055510.1 | - | 1 | 2263 |