Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268492.2 | XP_005268549.1 | 3 | 12974 | ||
XM_005268493.2 | XP_005268550.1 | 3 | 12370 | |||
XM_006714789.2 | XP_006714852.1 | 3 | 13034 | |||
XM_011537671.2 | XP_011535973.1 | 5 | 12869 | |||
GenBank mRNA | AK091723.1 | - | 1 | 263 | ||
BC036491.1 | - | 1 | 2080 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ942276.1 | - | 1 | 701 | Chr. 5 - 144484081 - 144484925 [+] on Build GRCh38 |