Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF116641.1 | - | 1 | 960 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ354065.1 | - | 1 | 194 | Chr. 14 - 58288798 - 58289120 [+] on Build GRCh38 |