Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120496.1 | 4 | 761 | |||
NR_120497.1 | 3 | 649 | ||||
GenBank mRNA | AK128534.1 | - | 2 | 1583 |