Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020791.2 | NP_065842.1 | 20 | 10242 | ||
NM_025142.1 | NP_079418.1 | 18 | 9798 | |||
XM_011525060.2 | XP_011523362.1 | 20 | 10264 | |||
GenBank mRNA | AK025902.1 | - | 1 | 646 | ||
AL833649.1 | - | 1 | 480 | |||
BC024166.1 | - | 1 | 513 | |||
BC037535.2 | - | 1 | 654 | |||
BC037740.2 | - | 1 | 3179 | |||
BC047652.1 | - | 1 | 1081 | |||
BC051737.1 | - | 1 | 3010 |