Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020791.2 | NP_065842.1 | 20 | 11964 | ||
NM_025142.1 | NP_079418.1 | 18 | 11520 | |||
XM_011525060.2 | XP_011523362.1 | 20 | 11986 | |||
GenBank mRNA | AL110163.1 | - | 1 | 1414 | ||
AL833649.1 | - | 1 | 2205 | |||
BC024166.1 | - | 1 | 2235 | |||
BC037740.2 | - | 1 | 4904 | |||
BC047652.1 | - | 1 | 2806 | |||
BC051737.1 | - | 1 | 4735 |