Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020791.2 | NP_065842.1 | 20 | 8278 | ||
NM_025142.1 | NP_079418.1 | 18 | 7834 | |||
XM_011525060.2 | XP_011523362.1 | 20 | 8300 | |||
GenBank mRNA | BC037740.2 | - | 1 | 1214 | ||
BC051737.1 | - | 1 | 1048 |