Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MELTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005929.5 | NP_005920.2 | 16 | 3329 | ||
XM_006713643.3 | XP_006713706.2 | 16 | 3406 | |||
XM_011512850.2 | XP_011511152.1 | 16 | 3409 | |||
GenBank mRNA | BC034933.1 | - | 1 | 303 | ||
BC068085.1 | - | 1 | 75 | |||
BC082761.1 | - | 1 | 75 |