Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038397.2 | 1 | 5365 | |||
NR_103486.1 | 4 | 1459 | ||||
GenBank mRNA | AK021543.1 | - | 1 | 1119 | ||
FJ623959.1 | - | 1 | 5365 |