Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038397.2 | 1 | 4624 | |||
NR_103486.1 | 4 | 718 | ||||
GenBank mRNA | AK021543.1 | - | 1 | 378 | ||
FJ623959.1 | - | 1 | 4624 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AU117617.1 | - | 1 | 378 | Chr. 1 - 172139707 - 172140589 [-] on Build GRCh38 |