Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC71L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175884.4 | NP_787080.2 | 1 | 2795 | ||
GenBank mRNA | AK098422.1 | - | 1 | 564 |