Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024419.4 | NP_077733.3 | 10 | 2241 | ||
NR_110601.1 | 9 | 2090 | ||||
NR_110602.1 | 9 | 2175 | ||||
NR_111989.1 | 9 | 1993 | ||||
XM_011525487.2 | XP_011523789.1 | 10 | 2616 | |||
XM_011525488.2 | XP_011523790.1 | 9 | 2517 | |||
XM_011525489.2 | XP_011523791.1 | 9 | 6928 | |||
XM_017025357.1 | XP_016880846.1 | 9 | 7001 | |||
XM_017025358.1 | XP_016880847.1 | 10 | 6463 | |||
XM_017025359.1 | XP_016880848.1 | 9 | 6314 | |||
XM_017025360.1 | XP_016880849.1 | 9 | 6364 | |||
XM_017025361.1 | XP_016880850.1 | 9 | 6620 | |||
XM_017025362.1 | XP_016880851.1 | 10 | 6082 | |||
XM_017025363.1 | XP_016880852.1 | 10 | 2187 | |||
XM_017025364.1 | XP_016880853.1 | 8 | 6522 | |||
XM_017025365.1 | XP_016880854.1 | 9 | 2089 | |||
XM_017025366.1 | XP_016880855.1 | 9 | 2136 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK090407.1 | - | 1 | 1800 | Chr. 17 - 78422774 - 78427966 [+] on Build GRCh38 |