Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 23 | 6318 | ||
NR_125340.1 | 20 | 6065 | ||||
XM_005244931.2 | XP_005244988.1 | 22 | 6258 | |||
XM_006711192.2 | XP_006711255.1 | 24 | 6228 | |||
XM_006711193.3 | XP_006711256.1 | 24 | 6360 | |||
XM_006711194.3 | XP_006711257.1 | 11 | 16356 | |||
XM_011509246.2 | XP_011507548.1 | 23 | 6179 | |||
XM_011509247.1 | XP_011507549.1 | 22 | 6119 | |||
XM_011509248.2 | XP_011507550.1 | 23 | 6112 | |||
XM_011509249.2 | XP_011507551.1 | 24 | 6417 | |||
XM_017000470.1 | XP_016855959.1 | 24 | 6306 | |||
XM_017000471.1 | XP_016855960.1 | 23 | 6168 | |||
XM_017000472.1 | XP_016855961.1 | 19 | 5792 | |||
GenBank mRNA | AK093982.1 | - | 1 | 211 |