Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033651.1 | 3 | 3846 | |||
NR_033652.1 | 5 | 4441 | ||||
GenBank mRNA | BX647470.1 | - | 3 | 3881 |