Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124198.1 | - | 1 | 1017 |
Gene Symbol: PIGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007240.1 | XP_016862729.1 | 1 | 1017 | ||
XM_017007241.1 | XP_016862730.1 | 2 | 2394 |