Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006713250.3 | XP_006713313.1 | 9 | 6539 | ||
XM_006713251.3 | XP_006713314.1 | 9 | 6716 | |||
XM_006713252.3 | XP_006713315.1 | 8 | 6396 | |||
XM_011533938.2 | XP_011532240.1 | 9 | 6573 | |||
XM_011533939.2 | XP_011532241.1 | 9 | 7412 | |||
XM_017006797.1 | XP_016862286.1 | 9 | 6570 | |||
XM_017006798.1 | XP_016862287.1 | 9 | 6562 | |||
XM_017006799.1 | XP_016862288.1 | 8 | 6393 |