Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648063.1 | - | 2 | 764 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC022826.1 | - | 1 | 448 | Chr. 1 - 156110451 - 156111467 [+] on Build GRCh38 |