Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933432I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045657.1 | 3 | 935 | |||
GenBank mRNA | AK017023.1 | - | 3 | 936 |