Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031243.1 | - | 1 | 1458 | ||
AK042703.1 | - | 1 | 1878 | |||
AK086149.1 | - | 2 | 2056 |