Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6330418K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045821.1 | 2 | 129 | |||
GenBank mRNA | AK005306.1 | - | 2 | 105 | ||
AK031849.1 | - | 2 | 438 |