Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK149635.1 | - | 1 | 2237 | ||
AK149792.1 | - | 1 | 2237 | |||
BC031980.1 | - | 1 | 163 |